Learn More
Corning™ cmRL 1066
BIOPHARMA
Product Code. 15343561
Change view
Click to view available options
Quantity:
6 x 500 mL
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15343561 6 x 500 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15343561 Supplier Corning™ Supplier No. 15110CV
  • ONLINE EXCLUSIVE PRICE
    € 324.00
    € 312.95 / Pack of 6
    Save € 11.05
    3% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use in both serum supplemented and unsupplemented culture
(This is suggested replacement for 15-110-CVR.)
TRUSTED_SUSTAINABILITY
Specifications
Product Type cmRL 1066
Quantity 6 x 500 mL
pH 6.9 to 7.3
Content And Storage 2° to 8°C