Learn More
Corning™ cellgro™ L-glutamine Powder
An essential amino acid and a key component of culture media
Merk:  Corning™ 61-030-RO
Overige gegevens : Gewicht : 0.54000kg
 Bekijk meerdere types van dit product via de
Produktcode. 15333751
  • € 263.55 / 500 gram
Geschatte verzenddatum
bij leverancier 05-06-2018
Toevoegen aan winkelmandje
Beschrijving
Beschrijving
Cellgro incorporates many principles developed by the pharmaceutical industry including the use of WFI quality water, raw material testing, in-process monitoring, and finished product testing.
  • Serves as major energy source for propagating cells
  • Molecular Weight: 146.15
Specificatie
Specificatie
Powder
5 to 6
L-Glutamine
Plastic Bottle
500g
2° to 8°C
Video's