Learn More
Corning™ CellSTACK™ Stacker
BIOPHARMAGREENER_CHOICE
Product Code. 10051442 Shop All Corning Products
Click to view available options
Material:
Plastic
Unit Size:
Pack of 5
Product Code. 10051442
Brand: Corning™ 3331
Only null left
Add to basket
Add to basket
Material: Plastic; For Use With: Incubator
A stacking device models the top surface of the CellSTACK™ chamber, securely nesting in the strengthening ribs. It is used to optimize incubator space while providing clearance for venting
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Incubator
Material Plastic
Type Stacker
For Use With (Application) Incubator