Description
The Costar™ Transtar-96™ System is a portable, autoclavable liquid handling system designed for use with 96 well plates. A sterile 96 tip disposable cartridge, which loads into the Transtar-96™ system, enables liquids to be aspirated, transferred and dispensed over a volume range of 25 to 200 μL in 5 μL increments. -Ideal for changing cell culture media and screening monoclonal antibodies. -Accuracy is rated at +/- 5% at all volume levels. -For sterile replacement cartridges, see No. 7610. -For sterile replacement reservoirs, see Nos. 4876 (open), 4877 (12-channel), or 4878 (8-channel). Refer to accessories tab below.
Specifications
Specifications
| Product Type | Liquid Transfer System |
| Autoclavable | Autoclavable |
| Channels | 96 |
| For Use With (Application) | Designed for use with 96-well plates, changing cell culture media and screening monoclonal antibodies |
| Sterility | Sterile |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.