Learn More
Corning™ DeckWorks™ Tip Stations, Nonsterile
Designed for single and multichannel pipetting stations
Brand: Corning™ 4142
Additional Details : Weight : 3.62874kg
Description
Corning DeckWorks tip station maximizes bench space while minimizing plastic waste. Tip stations contain 960 tips, are available in extended-length 10 μL and traditional 200 μL sizes, and are the ideal single and multi-channel pipetting stations. The 5-layer deck design (2 x 96) matches the microtiter plate format. This stable pipetting platform fits easily into sterile hood environments. Lids can be snap-positioned to expose a single side (96 tips) only. Applications include pre- and post-PCR procedures, cell culture, and assays requiring large tip quantities. Corning DeckWorks tip stations are offered both sterile and nonsterile.
Specifications
1 to 200 μL | |
Natural | |
Multi-pack | |
Yes | |
Polypropylene |
Non-sterile | |
Round | |
Universal Fit, Multichannel Pipetting Stations | |
DeckWorks |