Learn More
Corning™ HYPERStack™ Accessories: Stacking Tray
Shop All Corning Products
Click to view available options
:
Stacking Tray
Specifications
Specifications
| Color | Clear |
| Material | ABS Plastic |
| For Use With (Equipment) | Hyperstack™ cell culture vessels |
| Product Type | Stacking Tray |