Learn More
Corning™ LSE™ Digital Dry Bath Accessory, Single Blocks
Product Code. p-4423314 Shop All Corning Products
Click to view available options
No. of Places:
5
6
12
20
24
Tube Size:
0.5 mL
10 mm
12 mm
13 mm
1.5 mL
16 mm
2 mL
25 mm
15 mL
50 mL
Unit Size:
Each
Product Code. 11373815
Brand: Corning™ 480117
Only null left
Add to basket
Add to basket
Choice of configurations for use with LSE Digital Dry Bath Heater
The single and dual block configurations are used in the Corning LSE Digital Dry Bath Heaters.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 2 mL
For Use With (Equipment) LSE Dry Bath Heater
Material Stainless Steel
No. of Places 24
Type Block
Tube Size 2 mL