Learn More
Corning™ LSE™ Vortex Mixer Accessory, Processing Heads
Heads allow for processing of larger numbers of tubes
€ 38.12 - € 47.11
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
11911981
|
Corning™
480100 |
€ 38.48
Each |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||||
11921981
|
Corning™
480101 |
24 x 1.5/2.0 mL tubes, 24 x 0.5 mL tubes and 32 x 0.2 mL tubes/4 tube strips |
€ 38.12
Each |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
11941981
|
Corning™
480102 |
1 Microplate, 64 x 0.2 mL tubes, 8 x 0.2 mL tube strips |
€ 47.11
Each |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
11931981
|
Corning™
480103 |
8 x 15 mL tubes, 8 x 12 mm dia. tubes, and 8 x 13 mm dia. tubes |
€ 47.11
Each |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
11951981
|
Corning™
480104 |
6 x 50 mL tubes |
€ 47.11
Each |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
Description
This is an optional head for processing larger numbers of tubes, for use with Corning™ LSETM Vortex Mixer.Specifications
LSE Vortex Mixer |