Learn More
Corning™ Universal Fit Pipet Tips: Racked, Nonsterile
GREENER_CHOICE
Product Code. 10651462 Shop All Corning Products
Click to view available options
Volume (Metric):
1 to 200 μL
100 to 1000 μL
Color:
Blue
Natural
Yellow
Unit Size:
Pack of 1000
Pack of 960
Product Code. 10651462
Brand: Corning™ 4865
Only null left
Add to basket
Add to basket
Designed to assure a reliable fit with major brand pipetters
  • Corning universal fit tips are designed to provide a reliable fit with most major brand pipettors. (A Pipet Tip Compatibility Guide can be requested or downloaded from the Corning website).
  • Beveled orifice helps ensure accurate fluid delivery.
  • 1-200 μL universal fit tips are graduated at the 250 μL, 500 μL, and 1,000 μL volumes.
  • Select from three packaging options: Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design. Bulk packed tips are nonsterile and economical.
  • RNase-/DNase-free.
  • Nonpyrogenic.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 1 to 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Yellow
Format Rack
Tip Style Round
Packaging Rack
For Use With (Equipment) Universal Fit
Graduated Yes
Material Polypropylene