missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CORO1B Antibody (OTI3D10), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72086C
This item is not returnable.
View return policy
Description
CORO1B Monoclonal specifically detects CORO1B in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| CORO1B | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CORO1B | |
| Full length recombinant protein of human CORO1B (NP_065174) produced in HEK293T cells. | |
| 0.1 mL | |
| 57175.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI3D10 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| coronin, actin binding protein, 1B, coronin, actin-binding protein, 1B, coronin-1B, coronin-2, DKFZp762I166 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction