Learn More
CORO1B Mouse anti-Human, Clone: OTI3D10, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF501573
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Members of the coronin family, such as CORO1B, are WD repeat-containing actin-binding proteins that regulate cell motility.Specifications
| CORO1B | |
| Monoclonal | |
| Unconjugated | |
| CORO1B | |
| CORONIN-2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 57175 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI3D10 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9BR76 | |
| CORO1B | |
| Full length human recombit protein of human CORO1B produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.