missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CORO7 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81790R
This item is not returnable.
View return policy
Description
CORO7 Polyclonal antibody specifically detects CORO7 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| CORO7 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 79585 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 550 | |
| coronin-7, 0610011B16Rik, coronin 7, CRN7, POD1 | |
| CORO7 antibody was raised against an 17 amino acid synthetic peptide near the carboxy terminus of human CORO7. The immunogen is located within the last 50 amino acids of CORO7. Amino Acid Squence: YLEEKSDQQKKEELLNAM | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction