missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CORO7 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18450FR
This item is not returnable.
View return policy
Description
CORO7 Polyclonal antibody specifically detects CORO7 in Human samples. It is validated for Western Blot
Specifications
| CORO7 | |
| Polyclonal | |
| Western Blot | |
| coronin-7, 0610011B16Rik, coronin 7, CRN7, POD1 | |
| Recognizes region between residues Between Between 400 and 450.. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 79585 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction