missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coronin-1a Antibody (OTI1A5), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71680MFV610
This item is not returnable.
View return policy
Description
Coronin-1a Monoclonal antibody specifically detects Coronin-1a in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| Coronin-1a | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| OTI1A5 | |
| Flow Cytometry | |
| clipin-A, CLIPINA, CORO1, coronin, actin binding protein, 1A, coronin, actin-binding protein, 1A, coronin-1, coronin-1A, Coronin-like protein A, Coronin-like protein p57, FLJ41407, HCORO1, MGC117380, p57, TACOCLABP, Tryptophan aspartate-containing coat protein | |
| Full length human recombinant protein of human CORO1A(NP_009005) produced in HEK293T cell. | |
| 0.1 mL | |
| Cell Cycle and Replication, Immunology, Innate Immunity, Neuroscience | |
| 11151 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction