Learn More
Coronin 2B Polyclonal Antibody
Coronin 2B Polyclonal Antibody
Brand:  Invitrogen PA569820
Product Code. 15808233
  • € 520.00 / 100µL
Description
Coronin 2B Polyclonal Antibody for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Coronin 2B
Polyclonal
Unconjugated
CORO2B
clipin C; CLIPINC; Clipin-C; CORO2B; coronin 2B; coronin, actin binding protein, 2B; coronin-2B; Coronin-like protein C; E130012P22; E130012P22Rik; KIAA0925; Protein FC96
Rabbit
Affinity Chromatography
RUO
235431
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q8BH44
CORO2B
synthetic peptide corresponding to a region of mouse Coronin 2B (aa 275-324).