missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cortactin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11381-1-AP-20UL
This item is not returnable.
View return policy
Description
RSK3 is a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 non-identical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Mutations in the gene have been associated with Coffin-Lowry syndrome (CLS).Specifications
| Cortactin | |
| Polyclonal | |
| Unconjugated | |
| CTTN | |
| Amplaxin, cortactin, CTTN, EMS1, Oncogene EMS1, Src substrate cortactin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13043, 2017, 60465 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Flow Cytometry | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q14247, Q60598, Q66HL2 | |
| Cttn | |
| Cortactin Fusion Protein Ag1940 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction