missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Cortisol Parameter Assay Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Description
The Cortisol Parameter Assay Kit quantifies multi-species Cortisol in cell culture supernates (10 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), saliva (20 ul), urine (10 ul).
Specifications
Specifications
| Quantity | 1 Kit |
| Species | Multi-species |
| Conjugate | Biotin |
| Binding Target | Cortisol |
| Assay Sensitivity | 0.111 ng/mL |
| Assay Range | 0.16 to 10 ng/mL |
| For Use With (Application) | ELISA |
| Sample Type | Cell Culture Supernates (10uL), Serum (10uL), EDTA Plasma (10uL), Heparin Plasma (10uL), Saliva (20uL), Urine (10uL) |
| Test Time | 2.5 hr. |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction