missing translation for 'onlineSavingsMsg'
Learn More

Corylin, MedChemExpress

Product Code. 16401358
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16401358

Brand: Medchem Express HYN0236

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Corylin is a major bioactive compound isolated from Psoralea corylifolia L; antibiotic or anticancer compound.

Corylin is a major bioactive compound isolated from Psoralea corylifolia L

Specifications

Chemical Name or Material Corylin
Physical Form Powder
CAS 53947-92-5
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C20H16O4
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 100 mg/mL (312.17 mM)
SMILES O=C1C(C2=CC=C3C(C=CC(C)(C)O3)=C2)=COC4=CC(O)=CC=C14
Molecular Weight (g/mol) 320.34
Formula Weight 320.34
Percent Purity 98.54%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.