Läs mer
Corning™ HTS Transwell™ Clear Untreated Reservoirs
GREENER_CHOICE
Produktkod. 10461242 Handla allt Corning Produkter
Klicka för att se tillgängliga alternativ
No. per Pack:
12/Pk.
Förpackningsstorlek:
Pack of 48
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 10461242
Brand: Corning™ 3395
Bara null kvar
Lägg i varukorgen
Lägg i varukorgen
Denna artikel kan inte returneras. Se returpolicy
Manufactured from clear plastic for maximum visibility. Corning™ HTS Transwell™ Clear Untreated Reservoir is for use with HTS Transwell Permeable Supports.
  • For use as reservoir tray with HTS Transwell Permeable Supports
  • Not Treated for cell attachment
  • Same footprint as standard 24 well plate but with single rectangular well
TRUSTED_SUSTAINABILITY
Specifikationer
Color Clear
Material Polystyrene
Sterility Sterile
Surface Treatment Non-treated
Type Cell Culture Insert System
For Use With (Equipment) HTS Transwell™ Permeable Supports
No. per Case 48/Cs.
No. per Pack 12/Pk.