Learn More
Corning™ Costar™ Netwell™ Carrier Kit, Non-Sterile
Shop All Corning Products
Click to view available options
Diameter (Metric) Outer:
15 mm
24 mm
Unit Size:
Pack of 8
Description
Specifications
Specifications
| Type | Carrier Kit |
| For Use With (Application) | Allows simultaneous processing of up to 6 samples per carrier |
| Diameter (Metric) Outer | 24 mm |
| No. per Pack | 8/Pk. |
| No. per Case | 8/Cs. |