Learn More
Corning™ Costar™ Netwell™ Carrier Kit, Non-Sterile
GREENER_CHOICE
Product Code. 10631272 Shop All Corning Products
Click to view available options
Diameter (Metric) Outer:
15 mm
24 mm
Unit Size:
Pack of 8
Product Code. 10631272
Brand: Corning™ 3520
Only null left
Add to basket
Add to basket
Allows simultaneous processing
  • These specially designed Netwell carriers and handles allow simultaneous processing of up to 12 Netwell inserts per carrier
  • Each carrier kit contains eight carriers and eight handles
  • TRUSTED_SUSTAINABILITY
    Specifications
    Type Carrier Kit
    For Use With (Application) Allows simultaneous processing of up to (12) samples per carrier
    Diameter (Metric) Outer 15 mm
    No. per Pack 8/Pk.
    No. per Case 8/Cs.