missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-Costunolide, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | Costunolide |
| CAS | 553-21-9 |
| Molecular Formula | C15 H20 O2 |
| Quantity | 5 mg |
| Synonym | Cyclodeca[b]furan-2(3H)-one, 3a,4,5,8,9,11a-hexahydro-6,10-dimethyl-3-methylene-, (3aS,6E,10E,11aR)-, Costunolide (6CI), Cyclodeca[b]furan-2(3H)-one, 3a,4,5,8,9,11a-hexahydro-6,10-dimethyl-3-methylene-, [3aS-(3aR*,6E,10E,11aS*)]-, Germacra-1(10),4,11(13)-trien-12-oic acid, 6α-hydroxy-, γ-lactone, (E,E)- (8CI), (+)-Costunolide, (3aS,6E,10E,11aR)-3a,4,5,8,9,11a-Hexahydro-6,10-dimethyl-3-methylenecyclodeca[b]furan-2(3H)-one, Costunolid, Costus lactone, NSC 106404 |
| InChI Formula | InChI=1S/C15H20O2/c1-10-5-4-6-11(2)9-14-13(8-7-10)12(3)15(16)17-14/h5,9,13-14H,3-4,6-8H2,1-2H3/b10-5+,11-9+/t13-,14+/m0/s1 |
| SMILES | C\C\1=C/CC\C(=C\[C@H]2OC(=O)C(=C)[C@@H]2CC1)\C |
| IUPAC Name | (3aS,6E,10E,11aR)-6,10-dimethyl-3-methylidene-3a,4,5,8,9,11a-hexahydrocyclodeca[b]furan-2-one |
| Molecular Weight (g/mol) | 232.32 |
| Formula Weight | 232.1463 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction