missing translation for 'onlineSavingsMsg'
Learn More

Cot inhibitor-1, MedChemExpress

Product Code. 16483736
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16483736

Brand: Medchem Express HY32015

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cot inhibitor-1 (compound 28) is a selective tumor progression loci-2 (Tpl2) kinase inhibitor with an IC50 of 28 nM. Cot inhibitor-1 shows an inhibition of TNF-alpha production in human whole blood with an IC50 of 5.7 nM.

Cot inhibitor-1 (compound 28) is a selective tumor progression loci-2 (Tpl2) kinase inhibitor with an IC50 of 28 nM. Cot inhibitor-1 shows an inhibition of TNF-alpha production in human whole blood with an IC50 of 5.7 nM.

Specifications

Chemical Name or Material Cot inhibitor-1
CAS 915365-57-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C27H27Cl2FN8
Quantity 25 mg
Solubility Information DMSO : 25 mg/mL (45.17 mM; Need ultrasonic)
SMILES N#CC1=C(C2=CC(NCC3=CN(CCN4CCCCCC4)N=N3)=CC(Cl)=C2N=C1)NC5=CC=C(F)C(Cl)=C5
Molecular Weight (g/mol) 553.46
Formula Weight 553.46
Percent Purity 95.03%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.