missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COTL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10781-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes one of the numerous actin-binding proteins which regulate the actin cytoskeleton. This protein binds F-actin, and also interacts with 5-lipoxygenase, which is the first committed enzyme in leukotriene biosynthesis. Although this gene has been reported to map to chromosome 17 in the Smith-Magenis syndrome region, the best alignments for this gene are to chromosome 16. The Smith-Magenis syndrome region is the site of two related pseudogenes.Specifications
| COTL1 | |
| Polyclonal | |
| Unconjugated | |
| COTL1 | |
| CLP, Coactosin like protein, COTL1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23406, 361422, 72042 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| B0BNA5, Q14019, Q9CQI6 | |
| Cotl1 | |
| COTL1 Fusion Protein Ag1230 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction