Learn More
COTL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10781-1-AP-20UL
 View more versions of this product
Product Code. 16837393
  • € 145.00 / 20µL
Description
This gene encodes one of the numerous actin-binding proteins which regulate the actin cytoskeleton. This protein binds F-actin, and also interacts with 5-lipoxygenase, which is the first committed enzyme in leukotriene biosynthesis. Although this gene has been reported to map to chromosome 17 in the Smith-Magenis syndrome region, the best alignments for this gene are to chromosome 16. The Smith-Magenis syndrome region is the site of two related pseudogenes.
Specifications
Specifications
COTL1
Polyclonal
Unconjugated
COTL1
CLP, Coactosin like protein, COTL1
Rabbit
Antigen Affinity Chromatography
RUO
23406, 361422, 72042
-20°C
Liquid
Immunocytochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence
0.35 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
B0BNA5, Q14019, Q9CQI6
Cotl1
COTL1 Fusion Protein Ag1230
20 μL
Primary
Rat, Human, Mouse