Learn More
ZVG™ Cotton Cleaning Cloths
Description
- 100% cotton
- Predominantly seamless
- Lint-free, absorbent material
Specifications
Specifications
| Product Type | Cleaning Cloth |
| Material | Cotton |
| Certifications/Compliance | DIN 61650 |
| Color | Multicolored |
| Disposable | Single-use |
| Packaging Quantity | 10 kg |
| Sterility | Non-sterile |