missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cotton Rat IL-1 alpha/IL-1F1 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF552
This item is not returnable.
View return policy
Description
IL-1 alpha/IL-1F1 Polyclonal specifically detects IL-1 alpha/IL-1F1 in Cotton Rat samples. It is validated for Western Blot.
Specifications
| IL-1 alpha/IL-1F1 | |
| Polyclonal | |
| Biotin | |
| BAF, Hematopoietin-1, IL1, IL1 alpha, IL-1 alpha, IL1A, IL-1A, IL1-ALPHA, IL1F1, IL-1F1, IL1F1hematopoietin-1, interleukin 1, alpha, interleukin-1 alpha, LAF, LEM, preinterleukin 1 alpha, pro-interleukin-1-alpha | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3552 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| IL1A | |
| E. coli-derived recombinant cotton rat IL-1 alpha/IL-1F1 | |
| 50 μg | |
| Primary | |
| Detects cotton rat IL-1 alpha/IL-1F1 in Western blots. In Western blots, approximately 40% cross-reactivity with recombinant rat IL-1 alpha is observed, 20% cross-reactivity with recombinant mouse IL-1 alpha is observed, and less than 1% cross-reactivity with recombinant porcine IL-1 alpha and recombinant human IL-1 alpha is observed. | |
| Cotton Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction