missing translation for 'onlineSavingsMsg'
Learn More

Coulter™ IMMUNO-TROL™ Control Cells

Product Code. 15498214
Encompass
Change view
Click to view available options
Quantity:
3 mL
Unit Size:
60 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15498214 3 mL 60 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15498214 Supplier Coulter™ Supplier No. 6607077

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Positive process control for flow cytometry

Specifications

Certifications/Compliance IVD, CE, FCM
Format Liquid
Includes Two 3mL vials of cells
No. of Tests 60
Quantity 3 mL
Product Type Immuno-trol Control Cells
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.