Learn More
Hirschmann™ Counting Chambers Double Net Ruling
Description
- Double net ruling
- Including 2 cover glasses for hemacytometer
- CE acc. IVD 98/79 EC
Certifications
CE acc. IVD 98/79 EC
Specifications
Specifications
| Product Type | Counting chambers, double net ruling |
| Description | Counting chamber acc. to Nageotte |
| Quantity | 1 Each |
| Depth (Metric) | 0.5 mm |
| Includes | 2 cover glasses for haemacytometer |