Learn More
Karl Hecht™ Assistent™ Circular Cover Glasses for Microscopy
GREENER_CHOICE
Product Code. 10545182
Click to view available options
Diameter (Metric):
15 mm
Unit Size:
Pack of 100
Product Code. 10545182
Brand: Karl Hecht™ 1001/0015
Only null left
Add to basket
Add to basket
Preserve and protect specimens on slides. Karl Hecht™ Assistent™ Circular Cover Glasses for Microscopy are manufactured from a super-transparent borosilicate glass, preventing virtually any color distortion.
  • 0.13 - 0.16mm thickness
  • Excellent flatness
  • With CE marking
  • Optimum resistance to chemical attack
  • Refractive index finely adapted to microscopes
  • High spectral transmission
  • Excellent internal glass quality with very low levels of inclusions, striae, bubbles, streaks, etc.
  • In boxes of 100 pieces, 10 boxes in a carton
Specifications
Product Type Coverslip
Shape Circle
Material Glass
Thickness (Metric) #1 (0.13 to 0.16 mm)
Diameter (Metric) 15 mm
Certifications CE
Quantity 100/Pk.