missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ratiolab™ Covers for 96-Well Micro-Racks
Click to view available options
Format:
8-well cover stripe
96-well cover mat
Unit Size:
Pack of 100
Pack of 1000
Description
- For use with 1.2 mL tube racks
- Available as an 8-well cover stripe or 96-well cover mat
- Accessory item
Specifications
Specifications
| Material | TPE |
| Format | 96-well cover mat |
| Product Type | Cover |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction