missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX-2 Antibody (COX2/1941), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08272AF594
This item is not returnable.
View return policy
Description
COX-2 Monoclonal antibody specifically detects COX-2 in Human samples. It is validated for Western Blot, ELISA
Specifications
| COX-2 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| COX-2, COX2cyclooxygenase 2b, cyclooxygenase-2, EC 1.14.99, EC 1.14.99.1, GRIPGHS, hCox-2, PGG/HS, PGH synthase 2, PGHS-2, PHS II, PHS-2, prostaglandin G/H synthase 2, prostaglandin G/H synthase and cyclooxygenase, Prostaglandin H2 synthase 2, Prostaglandin-endoperoxide synthase 2, prostaglandin-endoperoxide synthase 2 (prostaglandin G/H synthase andcyclooxygenase) | |
| Recombinant human COX-2 protein fragment (around aa 442-572) (exact sequence is proprietary) (Uniprot: P35354) | |
| 100 μg | |
| Cancer, Cell Biology, Cellular Markers, Cytokine Research, Hypoxia, Lipid and Metabolism, Signal Transduction | |
| 5743 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, ELISA | |
| COX2/1941 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction