missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COX10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA524207
This item is not returnable.
View return policy
Description
COX10 Polyclonal Antibody for Western Blot, IHC (P), Flow
Defects in COX10 are a cause of mitochondrial complex IV deficiency (MT-C4D); also known as cytochrome c oxidase deficiency. A disorder of the mitochondrial respiratory chain with heterogeneous clinical manifestations, ranging from isolated myopathy to severe multisystem disease affecting several tissues and organs. Features include hypertrophic cardiomyopathy, hepatomegaly and liver dysfunction, hypotonia, muscle weakness, excercise intolerance, developmental delay, delayed motor development and mental retardation. A subset of patients manifest Leigh syndrome.
Specifications
| COX10 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q12887 | |
| COX10 | |
| KLH conjugated synthetic peptide between 383-410 amino acids from the C-terminal region of human COX10. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| COX10 | |
| 2410004F01Rik; AU042636; COX10; COX10 heme A:farnesyltransferase cytochrome c oxidase assembly factor; COX10 homolog, cytochrome c oxidase assembly protein, heme A: farnesyltransferase; COX10, heme A:farnesyltransferase cytochrome c oxidase assembly factor; cytochrome c oxidase assembly homolog 10; cytochrome c oxidase assembly protein; cytochrome c oxidase assembly protein 10; cytochrome c oxidase subunit X; heme A: farnesyltransferase; heme O synthase; LOC691853; Protoheme IX farnesyltransferase, mitochondrial; similar to COX10 homolog, cytochrome c oxidase assembly protein, heme A: farnesyltransferase | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 1352 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction