missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COX14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560781
This item is not returnable.
View return policy
Description
Immunogen sequence: VRVYHYFQWR RAQRQAAEEQ KTSGIM Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 73%.
Core component of the MITRAC (mitochondrial translation regulation assembly intermediate of cytochrome c oxidase complex) complex, that regulates cytochrome c oxidase assembly. Requires for coordination of the early steps of cytochrome c oxidase assembly with the synthesis of MT-CO1.
Specifications
| COX14 | |
| Polyclonal | |
| Unconjugated | |
| COX14 | |
| C12orf62; COX14; COX14 cytochrome c oxidase assembly factor; COX14 cytochrome c oxidase assembly homolog; COX14, cytochrome c oxidase assembly factor; cytochrome c oxidase assembly homolog 14; cytochrome c oxidase assembly protein COX14 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84987 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96I36 | |
| COX14 | |
| Recombinant protein corresponding to Human COX14. Recombinant protein control fragment (Product #RP-97854). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction