missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX17 Monoclonal Antibody (OTI7H5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812124S
This item is not returnable.
View return policy
Description
COX17 Monoclonal antibody specifically detects COX17 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| COX17 | |
| Monoclonal | |
| 0.5 to 1.0 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P34931 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3305 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI7H5 | |
| Unconjugated | |
| Hspa1l | |
| AI037035; Cox17; COX17 cytochrome c oxidase assembly homolog; COX17 cytochrome c oxidase copper chaperone; COX17 homolog, cytochrome c oxidase assembly protein; COX17, cytochrome c oxidase copper chaperone; Cox17a; cytochrome c oxidase 17 copper chaperone; cytochrome c oxidase assembly homolog 17; cytochrome c oxidase assembly protein 17; cytochrome c oxidase copper chaperone; cytochrome c oxidase subunit XVII assembly protein homolog; cytochrome c oxidase, subunit XVII assembly protein homolog; human homolog of yeast mitochondrial copper recruitment | |
| Human recombinant protein fragment corresponding to amino acids 400-641 of human HSPA1L produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction