missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 188.00 - € 463.00
Specifications
| Antigen | COX3 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18608288
|
Novus Biologicals
NBP3-05581-100ul |
100 μg |
€ 463.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18607427
|
Novus Biologicals
NBP3-05581-20ul |
20 μg |
€ 188.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
COX3 Polyclonal antibody specifically detects COX3 in Mouse, Rat samples. It is validated for Western BlotSpecifications
| COX3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cardiovascular Biology, Cell Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 4514 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| COIII, COX3, MTCO3, MT-CO3, MT-CO3 mitochondrially encoded cytochrome c oxidase III | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of mouse COX3 (NP_904334.1). MTHQTHAYHMVNPSPWPLTGAFSALLLTSGLVMWFHYNSITLLTLGLLTNILTMYQWWRDVIREGTYQGHHTPIVQKGLRYGMILFIVSEVFFFAGFFWA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title