missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX4-I1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 225.00 - € 305.00
Specifications
| Antigen | COX4-I1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
COX4-I1 Polyclonal antibody specifically detects COX4-I1 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
| COX4-I1 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human COX4-I1. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title