missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COX4I2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116399
This item is not returnable.
View return policy
Description
COX4I2 Polyclonal Antibody for Western Blot
Component of the cytochrome c oxidase, the last enzyme in the mitochondrial electron transport chain which drives oxidative phosphorylation. The respiratory chain contains 3 multisubunit complexes succinate dehydrogenase (complex II, CII), ubiquinol-cytochrome c oxidoreductase (cytochrome b-c1 complex, complex III, CIII) and cytochrome c oxidase (complex IV, CIV), that cooperate to transfer electrons derived from NADH and succinate to molecular oxygen, creating an electrochemical gradient over the inner membrane that drives transmembrane transport and the ATP synthase. Cytochrome c oxidase is the component of the respiratory chain that catalyzes the reduction of oxygen to water. Electrons originating from reduced cytochrome c in the intermembrane space (IMS) are transferred via the dinuclear copper A center (CU(A)) of subunit 2 and heme A of subunbit 1 to the active site in subunit 1, a binuclear center (BNC) formed by heme A3 and copper B (CU(B)). The BNC reduces molecular oxygen to 2 water molecules using 4 electrons from cytochrome c in the IMS and 4 protons from the mitochondrial matrix. [Uniprot].
Specifications
| COX4I2 | |
| Polyclonal | |
| Unconjugated | |
| COX4I2 | |
| AL024441; COX; cox iv; COX IV-1; Cox IV-2; cox4; COX4-1; Cox4-2; Cox4a; Cox4b; cox4i1; cox4i1.S; Cox4i2; COX4L2; COXIV; COXIV-1; CoxIV-2; Cytochrome c oxidase polypeptide IV; cytochrome c oxidase subunit 4 isoform 1, mitochondrial; Cytochrome c oxidase subunit 4 isoform 2, mitochondrial; cytochrome c oxidase subunit 4I1; cytochrome c oxidase subunit 4I2; cytochrome c oxidase subunit IV; cytochrome c oxidase subunit IV isoform 1; cytochrome c oxidase subunit IV isoform 1 precursor; cytochrome c oxidase subunit IV isoform 1 S homeolog; cytochrome c oxidase subunit IV isoform 2; cytochrome c oxidase subunit IV isoform 2 (lung); cytochrome c oxidase subunit IV precursor EC 1.9.3.1; cytochrome c oxidase subunit IV-like 2; cytochrome c oxidase, subunit 4a; cytochrome c oxidase, subunit 4b; cytochrome c oxidase, subunit IV; cytochrome c oxidase, subunit IVa; cytochrome c oxidase, subunit IVb; cytochrome c oxydase subunit 4; dJ857M17.2; FLJ23483; hypothetical protein MGC73355; IV-1; mg:bb02d03; MGC72016; XELAEV_18024950mg; zgc:110058; zgc:73355 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 84682, 84701 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.45 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q91W29, Q96KJ9 | |
| COX4I2 | |
| KLH conjugated synthetic peptide between 141-171 amino acids from human COXIV Isoform 2. | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction