missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX4NB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00354-50ul
This item is not returnable.
View return policy
Description
COX4NB Polyclonal antibody specifically detects COX4NB in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| COX4NB | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-4 uL/mg of lysate, Immunohistochemistry-Paraffin 1:1000-1:4000 | |
| C16orf2, C16orf4, chromosome 16 open reading frame 2, COX4 neighbor, COX4AL, FAM158Bchromosome 16 open reading frame 4, neighbor of COX4, NOC4family with sequence similarity 158, member B, Protein FAM158B | |
| Produced in rabbits immunized with purified, recombinant Human COX4NB (Accession#: O43402; Met1-Cys210) | |
| 50 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 10328 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction