missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX4NB Antibody (OTI1D6) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71595
This item is not returnable.
View return policy
Description
COX4NB Monoclonal specifically detects COX4NB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| COX4NB | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:2000, Immunohistochemistry 1:150 | |
| C16orf2, C16orf4, chromosome 16 open reading frame 2, COX4 neighbor, COX4AL, FAM158Bchromosome 16 open reading frame 4, neighbor of COX4, NOC4family with sequence similarity 158, member B, Protein FAM158B | |
| Mouse | |
| 23.6 kDa | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1D6 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| EMC8 | |
| Full length human recombinant protein of human COX4NB(NP_006058) produced in HEK293T cell. | |
| Affinity Purified | |
| Core ESC Like Genes, Stem Cell Markers | |
| 10328 | |
| Human, Mouse, Rat | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction