missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX4NB Monoclonal Antibody (OTI5G1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA507066S
This item is not returnable.
View return policy
Description
COX4NB Monoclonal antibody specifically detects COX4NB in Human samples. It is validated for Western BlotSpecifications
| COX4NB | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O43402 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10328 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI5G1 | |
| Unconjugated | |
| Emc8 | |
| C16orf2; C16orf4; COX4 neighbor; COX4AL; COX4NB; Emc8; ER membrane protein complex subunit 8; FAM158B; family with sequence similarity 158, member B; neighbor of COX4; NOC4; Protein FAM158B | |
| Full length human recombinant protein of human COX4NB produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction