Learn More
COX4NB Mouse anti-Human, Clone: OTI5G1, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF507066
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Expressed in liver, pancreas, heart, lung, kidney, brain, skeletal muscle, and placenta, Expression levels are highest in pancreas and moderate in heart, skeletal muscle, and placenta.Specifications
| COX4NB | |
| Monoclonal | |
| Unconjugated | |
| EMC8 | |
| C16orf2, C16orf4, COX4NB, FAM158B, NOC4 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10328 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI5G1 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O43402 | |
| EMC8 | |
| Full length human recombit protein of human COX4NB produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.