missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX6B1 Antibody (SR1049), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22471-100ul
This item is not returnable.
View return policy
Description
COX6B1 Monoclonal antibody specifically detects COX6B1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications
| COX6B1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| SR1049 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| COX VIb-1, COX6B, COXG, COXVIb1, cytochrome c oxidase subunit 6B1, cytochrome c oxidase subunit Vib, Cytochrome c oxidase subunit VIb isoform 1, cytochrome c oxidase subunit VIb polypeptide 1 (ubiquitous) | |
| A synthesized peptide derived from human COX6B1 (Uniprot #: P14854) | |
| 100 μg | |
| Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 1340 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction