missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ COX6B1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15863014
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15863014 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15863014 Supplier Invitrogen™ Supplier No. PA568027

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

COX6B1 Polyclonal Antibody for Western Blot, IHC, IHC (P)

Connects the two COX monomers into the physiological dimeric form.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX6B1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 30% glycerol, 0.87% NaCl and 0.01% sodium azide; pH 7.3
Gene COX6B1
Gene Accession No. P14854
Gene Alias 2010000G05Rik; COX VIb-1; Cox6b; COX6B1; COXG; COXVIb1; cytochrome c oxidase subunit 6B1; cytochrome c oxidase subunit VIb; Cytochrome c oxidase subunit VIb isoform 1; cytochrome c oxidase subunit VIb polypeptide 1 (ubiquitous); cytochrome c oxidase, subunit VIb polypeptide 1; LOC688869; similar to cytochrome c oxidase, subunit VIb polypeptide 1
Gene Symbols COX6B1
Host Species Rabbit
Immunogen Synthesized peptide derived from human COX6B1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1340
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.