missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COX7A2L Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551292
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total COX7A2L protein.
The cytochrome c oxidase (COX) family of proteins function as the final electron donor in the respiratory chain to drive a proton gradient across the inner mitochondrial membrane, ultimately resulting in the production of water. The mammalian COX apoenzyme is a dimer, with each monomer consisting of 13 subunits, some of which are mitochondrial and some of which are nuclear. COX7a2 (cytochrome c oxidase subunit VIIa polypeptide 2), also known as COX7AL or COX7AL1, is an 83 amino acid protein that localizes to the inner mitochondrial membrane and exists as a component of the COX complex, playing an important role in electron transport. COX7a2L (cytochrome c oxidase subunit 7A-related protein), also known as COX7AR or COX7RP, is an inner mitochondrial membrane protein that consists of 114 amino acids and is induced by estrogen.
Specifications
| COX7A2L | |
| Polyclonal | |
| Unconjugated | |
| Cox7a2l | |
| COX7A2L; COX7AR; COX7a-related protein; COX7RP; cytochrome c oxidase subunit 7A2 like; cytochrome c oxidase subunit 7A-related protein, mitochondrial; cytochrome c oxidase subunit VIIa polypeptide 2 like; cytochrome c oxidase subunit VIIa polypeptide 2-like; cytochrome c oxidase subunit VIIa-related protein; cytochrome c oxidase subunit VII-related protein; EB1; estrogen receptor binding CpG island; Scaf1; SIG81; SIG-81; Silg81; Silica-induced gene 81 protein; supercomplex assembly factor I | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9167 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.9 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O14548 | |
| Cox7a2l | |
| Synthetic peptide of human COX7A2L. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction