missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Coxsackievirus A6 VP2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5117350

Product Code. 17299843

  • € 497.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Centrifuge briefly prior to opening.

Coxsackievirus A6 (CVA6) is a human enterovirus associated with herpangina in infants. VP1, VP2, and VP3 all have a compact core structure consisting of a classical, beta-sandwich, jelly-roll fold, which forms the bulk of the capsid and each of these protein subunits also have several loops that decorate the outer surface of the capsid.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Coxsackievirus A6 VP2
Polyclonal
Unconjugated
Coxsackievirus
Recombinant protein encompassing a sequence within the center region of Coxsackievirus A6 VP2. The exact sequence is proprietary.
100 μL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Rabbit
Antigen affinity chromatography
RUO
Virus
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.