Learn More
Tocris Bioscience™ CP 690550 citrate
GREENER_CHOICE
Product Code. 30001514
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 30001514
Brand: Tocris Bioscience™ 4556/10
Potent JAK inhibitor
Potent JAK inhibitor (IC50 values are 1, 20, and 112 nM for JAK3, JAK2 and JAK1 respectively). Orally active immunosuppressant; exhibits efficacy in rodent rheumatoid arthritis models.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 540737-29-9
Molecular Formula C22H28N6O8
Specifications
Quantity 10 mg
Formula Weight 504.49
Purity >99%
Molecular Weight (g/mol) 504.5
Chemical Name or Material CP 690550 citrate