missing translation for 'onlineSavingsMsg'
Learn More

CP110 Rabbit anti-Canine, Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16880674 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16880674 150 μL 150µL
16890674 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16880674 Supplier Proteintech Supplier No. 127801AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GSG2 is required for normal alignment of chromosomes at metaphase.

Specifications

Antigen CP110
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CCP110
Gene Accession No. O43303, Q7TSH4
Gene Alias Centrosomal protein of 110 kDa, CP110, CP110 protein, KIAA0419
Gene Symbols Ccp110
Host Species Rabbit
Immunogen CP110 Fusion Protein Ag3489
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101565, 361634, 479826, 9738
Target Species Canine, Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.