missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPA1 (Carboxypeptidase A1) Monoclonal Antibody (OTI4E12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504606S
This item is not returnable.
View return policy
Description
CPA1 (Carboxypeptidase A1) Monoclonal antibody specifically detects CPA1 (Carboxypeptidase A1) in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| CPA1 (Carboxypeptidase A1) | |
| Monoclonal | |
| 0.58 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O43278 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6692 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI4E12 | |
| Unconjugated | |
| SPINT1 | |
| 0910001L12Rik; carboxypeptidase A; carboxypeptidase A1; carboxypeptidase A1 (pancreatic); carboxypeptidase A1, pancreatic; CPA; Cpa1; pancreatic carboxypeptidase A; pancreatic procarboxypeptidase A | |
| Full length human recombinant protein of human SPINT1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction