Learn More
CPA1 (Carboxypeptidase A1) Mouse anti-Human, Clone: OTI2A3, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF500053
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CPA1, also known as CPA, is a 419 amino acid secreted monomeric protein that is highly expressed in pancreatic tissue. Functioning as a pancreatic exopeptidase, CPA1 uses zinc as a cofactor to catalyze the release of C-terminal amino acids from a variety of proteins, thereby playing a key role in protein digestion and degradation. Via its catalytic activity, CPA1 is also thought to be involved in zymogen (proenzyme) inhibition, probably functioning to block enzyme activation pathways. Abnormal levels of CPA1 are associated with pancreatic cancer, suggesting a possible role in either tumor progression or tumor suppression events.Specifications
| CPA1 (Carboxypeptidase A1) | |
| Monoclonal | |
| Unconjugated | |
| CPA1 | |
| CPA | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1357 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2A3 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P15085 | |
| CPA1 | |
| Recombit protein expressed in E.coli corresponding to amino acids 1-419 of human CPA-1 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.